Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT2A Antibody (1055220), Biotin, Novus Biologicals™
SDP

Catalog No. NB297453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297453 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297453 Supplier Novus Biologicals Supplier No. FAB112501B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

5-HT2A Monoclonal specifically detects 5-HT2A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen 5-HT2A
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1055220
Conjugate Biotin
Dilution Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias 5-HT-2, 5-HT2 receptor, 5-HT-2A, 5-hydroxytryptamine (serotonin) receptor 2A, 5-hydroxytryptamine receptor 2A, HTR2,5-HT2A, serotonin 5-HT-2A receptor, Serotonin receptor 2A
Host Species Mouse
Immunogen Synthetic peptide corresponding to the amino acids 1-75 in the N-term extracellular domain of the Serotonin Receptor 5-HT2A (HTR2A)., Accession # P28223
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3356
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.