Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT2B Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB318301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318301 Supplier Novus Biologicals Supplier No. NBP298911PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-HT2B Polyclonal antibody specifically detects 5-HT2B in Human samples. It is validated for Western Blot

Specifications

Antigen 5-HT2B
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias 5-HT(2B), 5-HT-2B, 5-HT2B5-HT 2B receptor, 5-hydroxytryptamine (serotonin) receptor 2B, 5-hydroxytryptamine 2B receptor, 5-hydroxytryptamine receptor 2B, Serotonin receptor 2B
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human 5-HT2B.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3357
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.