Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT2C Antibody (496214) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB377598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377598 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377598 Supplier Novus Biologicals Supplier No. FAB5764W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

5-HT2C Monoclonal antibody specifically detects 5-HT2C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen 5-HT2C
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 496214
Conjugate DyLight 650
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 5-HT1C, 5-HT-1C, 5-HT-2C, 5-HT2C5-hydroxytryptamine receptor 1C, 5-HTR2C, 5-hydroxytryptamine (serotonin) receptor 2C, 5-hydroxytryptamine receptor 2C, HTR1C, serotonin 5-HT-2C receptor, Serotonin receptor 2C
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human 5HT2C
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3358
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.