Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-HT3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | 5-HT3A |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
5-HT3A Polyclonal antibody specifically detects 5-HT3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
5-HT3A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS (pH 7.2) and 40% Glycerol | |
3359 | |
IgG | |
Protein A purified |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
5-HT-3,5-HT3-A, 5-HT3R5-hydroxytryptamine receptor 3A, 5-hydroxytryptamine (serotonin) receptor 3A, 5-hydroxytryptamine receptor 3,5-HT3A5HT3R, HTR35HT3 serotonin receptor, Serotonin receptor 3A, Serotonin-gated ion channel receptor | |
This antibody was developed against a recombinant protein corresponding to amino acids: LRHLVLERIAWLLCLREQSTSQRPPATSQATKTDDCSAMGNHCSHMGGPQDFEKSPRDRCSPPPPPREASLAVCGLLQELSSIRQFLEKRDEI | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title