Learn More
Description
Specifications
Specifications
| Antigen | 5-HT3A |
| Applications | Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 3 ug/ml, Simple Western 1:100, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 5 ug/ml |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | P35563 |
| Gene Alias | 5-HT-3,5-HT3-A, 5-HT3R5-hydroxytryptamine receptor 3A, 5-hydroxytryptamine (serotonin) receptor 3A, 5-hydroxytryptamine receptor 3,5-HT3A5HT3R, HTR35HT3 serotonin receptor, Serotonin receptor 3A, Serotonin-gated ion channel receptor |
| Gene Symbols | HTR3A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
