Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT4 Antibody (631769), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB303004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303004 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303004 Supplier Novus Biologicals Supplier No. FAB6545MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

5-HT4 Monoclonal antibody specifically detects 5-HT4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry

Specifications

Antigen 5-HT4
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 631769
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias 5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, Serotonin receptor 4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human 5-HT4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, GPCR, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3360
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.