Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT4 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP178403R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178403R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178403R Supplier Novus Biologicals Supplier No. NBP178403R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-HT4 Polyclonal specifically detects 5-HT4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 5-HT4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q13639
Gene Alias 5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, Serotonin receptor 4
Gene Symbols HTR4
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human 5HT4 Receptor protein (between residues 350-388) [UniProt Q13639, isoform 1]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, GPCR, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3360
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.