Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146557 0.1 mL
NB018625 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP146557 Supplier Novus Biologicals Supplier No. NBP146557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

5-HT6 Polyclonal specifically detects 5-HT6 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen 5-HT6
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. P31388, Q9R1C8
Gene Alias 5-hydroxytryptamine (serotonin) receptor 6,5-HT65-HT6R, Serotonin receptor 6,5-HT-6,5-hydroxytryptamine receptor 6
Gene Symbols HTR6
Host Species Rabbit
Immunogen A synthetic peptide from aa region 400-450 of mouse 5HT6R conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3362
Test Specificity Specific for 5HT6R receptor.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.