Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT7 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056352T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056352T 0.025 mg
NB10056352 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056352T Supplier Novus Biologicals Supplier No. NB10056352SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

5-HT7 Polyclonal specifically detects 5-HT7 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Simple Western, Western Blot.

Specifications

Antigen 5-HT7
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Simple Western, Flow Cytometry reported in scientific literature (PMID 30602786), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (PMID 17940054)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P32305
Gene Alias 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7
Gene Symbols HTR7
Host Species Rabbit
Immunogen This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 13-28 of the rat 5-HT7R (AAA42134.1).
Molecular Weight of Antigen 54 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3363
Test Specificity The 5HT7 antibody recognizes all described 5HT7 receptor splice variants.
Target Species Canine, Mouse, Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.