Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT7 Antibody, Novus Biologicals™
SDP

Catalog No. NB018632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018632 0.025 mL
NBP146598 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018632 Supplier Novus Biologicals Supplier No. NBP1465980.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

5-HT7 Polyclonal specifically detects 5-HT7 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen 5-HT7
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1: 200 - 1: 2000, Immunohistochemistry 1: 200- 1: 2000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1: 200 - 1: 2000
Formulation Whole antisera with No Preservative
Gene Accession No. P34969
Gene Alias 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7
Gene Symbols HTR7
Host Species Rabbit
Immunogen A synthetic peptide from internal region of human 5HT7R receptor (Serotonin receptor 7R) conjugated to an immunogenic carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3363
Test Specificity Specific for 5HT7R receptor.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Mouse, Guinea Pig, Rat, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.