Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-HT7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$181.90 - $487.50
Specifications
Antigen | 5-HT7 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
5-HT7 Polyclonal specifically detects 5-HT7 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
5-HT7 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
P34969 | |
3363 | |
IgG | |
Unpurified | |
Specific for 5HT7R receptor. |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
Mouse, Guinea Pig, Rat, Human | |
5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7 | |
HTR7 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title