Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxynorbornane 2,6-Lactone 98.0+%, TCI America™
SDP

Catalog No. H147525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H147525G 25 g
H14755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H147525G Supplier TCI America Supplier No. H147525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92343-46-9
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD00988184
InChI Key SCDJKGVPVLZLED-UHFFFAOYSA-N
Synonym 6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one
PubChem CID 432639
SMILES C1C2CC3C1C(C2O)OC3=O

Specifications

Melting Point 158°C
Color Yellow
Quantity 25 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxynorbornane 2,6-Lactone
EINECSNumber (5)-6966
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.