Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 5-Iodotubercidin
GREENER_CHOICESDP

Catalog No. 17451 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-451 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-451 Supplier R&D Systems™ Supplier No. 1745/1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent adenosine kinase inhibitor; also inhibits nucleoside transporters and a range of other kinases

Potent adenosine kinase inhibitor (IC50 = 26 nM). Also nucleoside transporter inhibitor (IC50 values are < 25 nM, 7 μM and 15 μM for inhibition of [3H]adenosine, [3H]uridine and [3H]formycin B uptake respectively). Strongly stimulates glycogen synthesis in hepatocytes via activation of glycogen synthase. Also inhibits CK1, insulin receptor tyrosine kinase, phosphorylase kinase, PKA, CK2 and PKC (IC50 values are 0.4, 3.5, 5-10, 5-10, 10.9 and 27.7 μM respectively). Decreases hippocampal DNA methylation through adenosine kinase inhibition in vivo.

Chemical Identifiers

CAS 24386-93-4
Molecular Formula C11H13IN4O4

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 392.15
Purity 97%
Molecular Weight (g/mol) 392.15
Chemical Name or Material 5-Iodotubercidin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.