Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Lipoxygenase Antibody (1D4), Novus Biologicals™
SDP

Catalog No. NB165510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165510 Supplier Novus Biologicals Supplier No. NBP315171
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

5-Lipoxygenase Monoclonal antibody specifically detects 5-Lipoxygenase in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen 5-Lipoxygenase
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1D4
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias 5-lipoxygenase, 5-LO, 5-LOX, 5LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204
Host Species Rabbit
Immunogen A synthesized peptide derived from human 5-Lipoxygenase. UniProt ID (P09917).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 240
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.