Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Lipoxygenase Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1108748A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1108748A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1108748A32 Supplier Novus Biologicals Supplier No. NB11058748AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-Lipoxygenase Polyclonal specifically detects 5-Lipoxygenase in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 5-Lipoxygenase
Applications KnockDown, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 5-lipoxygenase, 5-LO, 5-LOX, 5LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204
Gene Symbols ALOX5
Host Species Rabbit
Immunogen A synthetic peptide made to a region within residues 100-200 of human 5-Lipoxygenase. [Swiss-ProtNo. P09917]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 240
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.