Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Lipoxygenase Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339213 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339213 Supplier Novus Biologicals Supplier No. NB11058748MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-Lipoxygenase Polyclonal antibody specifically detects 5-Lipoxygenase in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunocytochemistry/ Immunofluorescence, KnockDown

Specifications

Antigen 5-Lipoxygenase
Applications Western Blot, Immunocytochemistry, KnockDown
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias 5-lipoxygenase, 5-LO, 5-LOX, 5LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204
Host Species Rabbit
Immunogen A synthetic peptide made to a region within residues 100-200 of human 5-Lipoxygenase. [Swiss-Prot# P09917]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 240
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.