Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxysalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. M125425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M125425G 25 g
M12545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M125425G Supplier TCI America Supplier No. M125425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2612-02-4
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002459
InChI Key IZZIWIAOVZOBLF-UHFFFAOYSA-N
Synonym 5-methoxysalicylic acid, benzoic acid, 2-hydroxy-5-methoxy, 5-methoxy-2-hydroxybenzoic acid, 6-hydroxy-m-anisic acid, m-anisic acid, 6-hydroxy, 2-hydroxy-5-methoxybenzadelhyde, acid5-methoxysalicylic, 2-hydroxy-5-methoxybenzoic, 5-o-methyl gentisic acid, 2-hydroxy-5-methoxy-benzoic acid
PubChem CID 75787
IUPAC Name 2-hydroxy-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 25 g
Formula Weight 168.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxysalicylic Acid
EINECSNumber (3)-1647
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.