Learn More
Description
Ensure accurate, reproducible results in Dot Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunohistochemistry
Specifications
Specifications
| Antigen | 5-MethylCytosine |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, Immunoassay |
| Classification | Monoclonal |
| Clone | 4111 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen, Dot Blot 1:100 - 1:2000, Electron Microscopy, Methylated DNA Immunoprecipitation, Immunohistochemistry 1:50-1:500 |
| Formulation | PBS with No Preservative |
| Gene Alias | 5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl) |
| Host Species | Mouse |
| Immunogen | The immunogen used to generate this antibody corresponds to 5-Methylcytosine/ 5-mC |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
