Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-MethylCytosine Antibody, Novus Biologicals™
SDP

Catalog No. NB100744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100744 0.125 mg
NB017813 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100744 Supplier Novus Biologicals Supplier No. NB100744
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 3 publications

5-MethylCytosine Polyclonal specifically detects 5-MethylCytosine in All species, Human, Mouse, Rat samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen 5-MethylCytosine
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 3.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 25 to 100 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS (pH 7.5) with No Preservative
Gene Alias 5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl)
Host Species Sheep
Immunogen Chemical Small Molecule conjugated to KLH.
Purification Method Protein G purified
Quantity 0.125 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is specific for 5-Methyl Cytosine. It was tested using immunoprecipitation against 5-Methyl Cytosine and bromodeoxyuridine (BrdU) or a control (no antigen).
Target Species All species, Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.