Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-MethylCytosine Antibody, Novus Biologicals™

Sheep Polyclonal Antibody has been used in 3 publications
$192.47 - $443.24
Specifications
| Antigen | 5-MethylCytosine |
|---|---|
| Concentration | 3.0 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
5-MethylCytosine Polyclonal specifically detects 5-MethylCytosine in Human, Rat, All Species samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| 5-MethylCytosine | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.5) with No Preservative | |
| Chemical Small Molecule conjugated to KLH. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 3.0 mg/mL | |
| Unconjugated | |
| Sheep | |
| All species, Rat | |
| 5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl) | |
| IgG | |
| Protein G purified | |
| This is specific for 5-Methyl Cytosine. It was tested using immunoprecipitation against 5-Methyl Cytosine and bromodeoxyuridine (BrdU) or a control (no antigen). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title