Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-MethylCytosine Sheep anti-All, Rat, PE, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB100744PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100744PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100744PE Supplier Novus Biologicals Supplier No. NB100744PE

Sheep Polyclonal Antibody

5-MethylCytosine Polyclonal specifically detects 5-MethylCytosine in Human, Rat, All Species samples. It is validated for Flow Cytometry.

Specifications

Antigen 5-MethylCytosine
Applications Flow Cytometry
Classification Polyclonal
Conjugate PE
Dilution Flow Cytometry
Formulation PBS (pH 7.5) with No Preservative
Gene Alias 5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl)
Host Species Sheep
Immunogen Chemical Small Molecule conjugated to KLH.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is specific for 5-Methyl Cytosine. It was tested using immunoprecipitation against 5-Methyl Cytosine and bromodeoxyuridine (BrdU) or a control (no antigen).
Target Species Rat, All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.