Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitropyridine-2-carboxylic acid, 97+%

Catalog No. AAH6449903
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 30651-24-2 | C6H4N2O4 | 168.11 g/mol

It plays an important role in synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role in synthetic intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30651-24-2
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD04114182
InChI Key QKYRCTVBMNXTBT-UHFFFAOYSA-N
Synonym 5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid
PubChem CID 520488
IUPAC Name 5-nitropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 208°C to 212°C
Quantity 1 g
Beilstein 139209
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 168.11
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 5-Nitropyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.