Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Nucleotidase/CD73 Antibody (102), HRP, Novus Biologicals™
SDP

Catalog No. NB067710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067710 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067710 Supplier Novus Biologicals Supplier No. NBP289706H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for ELISA.

Specifications

Antigen 5'-Nucleotidase/CD73
Applications ELISA
Classification Monoclonal
Clone 102
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human 5'-Nucleotidase/CD73 (NP_002517.1; Met1-Lys547).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 4907
Test Specificity No cross-reactivity in ELISA with: Mouse NT5E
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.