Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Nucleotidase/CD73 Antibody (45M4F9) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP225237SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225237SS 0.025 mg
NBP225237 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225237SS Supplier Novus Biologicals Supplier No. NBP225237SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen 5'-Nucleotidase/CD73
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 45M4F9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Gene Accession No. NP_001191742
Gene Alias 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Gene Symbols NT5E
Host Species Mouse
Immunogen A portion of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4907
Test Specificity This product can detect Isoform 1 and Isoform of Human CD73.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.