Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5'-Nucleotidase/CD73 Antibody (45M4F9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $426.00
Specifications
| Antigen | 5'-Nucleotidase/CD73 |
|---|---|
| Clone | 45M4F9 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225237SS
![]() |
Novus Biologicals
NBP225237SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP225237
![]() |
Novus Biologicals
NBP225237 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| 5'-Nucleotidase/CD73 | |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| NP_001191742 | |
| 4907 | |
| A portion of amino acids 169-324 of human CD73 were used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 45M4F9 | |
| Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
| NT5E | |
| IgG2b κ | |
| Protein G purified | |
| This product can detect Isoform 1 and Isoform of Human CD73. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title