Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Nucleotidase/CD73 Antibody (496406), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB370000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370000 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370000 Supplier Novus Biologicals Supplier No. FAB4488K
Add to Cart
Add to Cart

Rat Monoclonal Antibody

5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen 5'-Nucleotidase/CD73
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 496406
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse 5'-Nucleotidase/CD73, Trp29-Lys549, Accession # Q61503
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 4907
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.