Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Nucleotidase/CD73 Antibody (606112) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB377554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377554 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377554 Supplier Novus Biologicals Supplier No. FAB5795W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

5'-Nucleotidase/CD73 Monoclonal antibody specifically detects 5'-Nucleotidase/CD73 in Human, Equine samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen 5'-Nucleotidase/CD73
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 606112
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human 5'-Nucleotidase/CD73, Trp27-Ile511, Accession # P21589
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 4907
Target Species Human, Equine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.