Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek 50 TS Microplate Washer, IVD PROMO

Catalog No. 50TS8VSIW25 Shop All Agilent Technologies Inc. Products
Click to view available options
Model:
50TS12
50TS12V
50TS16
50TS16V
50TS8
50TS8F
50TS8M
50TS8MF
50TS8V
This item is not returnable. View return policy

Catalog No. 50-TS8-VSIW25

Supplier: Agilent BioTek 50TS8VSI3044

This item is not returnable. View return policy

Offers excellent microplate washing performance. For In Vitro Diagnostic Use.

The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well

The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE and TUV marked. RoHS Compliant.
Connections USB (1)
Bottles Optional (2 to 4 L)
Plate Type 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Soak Time Programmable in minutes and seconds, up to 30 min.
Type Microplate Washer
Weight (Metric) 9.8 kg
Wash Cycles 1 to 10
Wash Volume 25 to 3000 μL/well
Display 4.3 in. LCD Color Touch Screen
Flow Rate 150 to 1000 μL/well/sec. (Manifold Dependent)
Fluid Delivery Type One Positive Displacement Syringe Drive
Model 50TS8V
Includes 96-well Plates only, Buffer Switching
Automation Interface 4.3 in. Color LCD Touch Screen Display
Weight (English) 22 lb.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.