Learn More
Agilent BioTek 50 TS Microplate Washer PROMO
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.

Specifications
Specifications
Certifications/Compliance | CE and TUV marked. RoHS Compliant. |
Connections | USB (1) |
Bottles | Optional (2 to 4 L) |
Plate Type | 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) |
Soak Time | Programmable in minutes and seconds, up to 30 min. |
Type | Microplate Washer |
Weight (Metric) | 9.8 kg |
Wash Cycles | 1 to 10 |
Wash Volume | 25 to 3000 μL/well |
Display | 4.3 in. LCD Color Touch Screen |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.