Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1 Antibody (1285A), Alexa Fluor 350, Novus Biologicals™
SDP

Catalog No. MB18771A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB18771A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB18771A350 Supplier Novus Biologicals Supplier No. MAB18771AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

53BP1 Monoclonal specifically detects 53BP1 in Human samples. It is validated for Immunocytochemistry, Western Blot.

Specifications

Antigen 53BP1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1285A
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen Recombinant monoclonal 53BP1 Antibody (1285A) was made to amino acids 350 - 400 of human 53BP1 (NP_005648.1).
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Protein Phosphatase, Tumor Suppressors, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 7158.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.