Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1 Antibody (1285C) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP254753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254753 0.1 mg
NBP254753SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254753 Supplier Novus Biologicals Supplier No. NBP254753
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

53BP1 Monoclonal specifically detects 53BP1 in Human, Mouse samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen 53BP1
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1285C
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Flow Cytometry 2.5-5 ug/ml, Immunohistochemistry 1.0 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1.0 ug/ml, Flow (Intracellular) 1 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen 53BP1 Antibody (1285C) was made to a synthetic peptide made to the C-terminal portion of human 53BP1 protein (between amino acids 1900-1972) [UniProt Q12888]
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Protein Phosphatase, Tumor Suppressors, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 7158
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.