Learn More
Description
Specifications
Specifications
| Antigen | 53BP1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), KnockDown, Proximity Ligation Assay, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 |
| Gene Symbols | TP53BP1 |
| Host Species | Rabbit |
| Immunogen | 53BP1 Antibody was made to a synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19 [Uniprot: Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
