Learn More
Description
Specifications
Specifications
| Antigen | 53BP1 |
| Applications | Chemotaxis, ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), KnockDown, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Chromatin Immunoprecipitation reported in scientific literature, Flow Cytometry 2-5 ug/million cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen reported in scientific literature, Chromatin Immunoprecipitation (ChIP), KnockDown Validated |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q12888 |
| Gene Alias | 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 |
| Gene Symbols | TP53BP1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
