Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100904 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100904 0.1 mL
NB100904SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100904 Supplier Novus Biologicals Supplier No. NB100904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 96 publications

53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse samples. It is validated for Block/Neutralize, Flow Cytometry, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, In-situ Hybridization, KnockDown, Proximity Ligation Assay, Western Blot.

Specifications

Antigen 53BP1
Applications Blocking Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), KnockDown, Proximity Ligation Assay, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:10000, Flow Cytometry 2 - 5 ug/mL, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 25512560), Immunohistochemistry-Paraffin 1:400, Proximity Ligation Assay reported in scientific literature (PMID 26734725), In-situ Hybridization reported in scientific literature (PMID 25407517), KnockDown Validated reported in scientific literature (PMID 31635359)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q12888
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen 53BP1 Antibody was made to a synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19 [Uniprot: Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7158
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Target Species Human, Mouse, Mouse
Content And Storage Store at -20 degreesC.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.