Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB100304UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100304UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100304UV Supplier Novus Biologicals Supplier No. NB100304UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

53BP1 Polyclonal specifically detects 53BP1 in Bat, Bovine, Canine, Fish, Goat, Human, Mouse, Primate, Rat samples. It is validated for ChIP Assay, Flow Cytometry, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In-situ Hybridization, KnockDown, Western Blot.

Specifications

Antigen 53BP1
Applications ChIP Assay, Flow Cytometry, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), KnockDown, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), Knockout Validated, Knockdown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of human 53BP1 (between amino acids 350-400) [NP_005648.1].
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7158
Target Species Bat, Bovine, Canine, Fish, Goat, Human, Mouse, Primate, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.