Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002858 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002858 Supplier Novus Biologicals Supplier No. NB1002858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

53BP2 Polyclonal specifically detects 53BP2 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen 53BP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_001026855.1
Gene Alias apoptosis-stimulating protein of p53, 2, ASPP2P53BP2,53BP2apoptosis-stimulating of p53 protein 2, Bbp, Bcl2-binding protein, p53-binding protein 2, p53BP2, PPP1R13A, Renal carcinoma antigen NY-REN-51, tumor protein p53 binding protein, 2, tumor protein p53-binding protein, 2, Tumor suppressor p53-binding protein 2
Gene Symbols TP53BP2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 425 and 475 of human Apoptosis-Stimulating Protein of p53, 2 (53BP2) using the numbering given in entry NP_001026855.1 (GeneID 7159).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7159
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.