Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyoxalase I Antibody (Glo1a), Novus Biologicals™
SDP

Catalog No. NBP119015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119015 0.1 mL
NBP119015SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP119015 Supplier Novus Biologicals Supplier No. NBP119015
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 5 publications

Glyoxalase I Monoclonal specifically detects Glyoxalase I in Human, Mouse samples. It is validated for Western Blot, Simple Western, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Glyoxalase I
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Glo1a
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Simple Western 1:50, ELISA 0.15 ng/ 1 ug protein, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature (10.3892/ol.2021.12808)
Gene Accession No. Q04760
Gene Alias Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase
Gene Symbols GLO1
Host Species Mouse
Immunogen Full length human GLO1 protein [Swiss-Prot #Q04760].
Molecular Weight of Antigen 21 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2739
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.