Learn More
Description
Specifications
Specifications
| Antigen | Glyoxalase I |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | Glo1a |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Simple Western 1:50, ELISA 0.15 ng/ 1 ug protein, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature (10.3892/ol.2021.12808) |
| Gene Accession No. | Q04760 |
| Gene Alias | Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase |
| Gene Symbols | GLO1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
