Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

58K Golgi Protein Antibody (FTCD/357), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB157654 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157654 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157654 Supplier Novus Biologicals Supplier No. NBP314274C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

58K Golgi Protein Monoclonal antibody specifically detects 58K Golgi Protein in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen 58K Golgi Protein
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone FTCD/357
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1
Host Species Mouse
Immunogen 58K Golgi Protein Protein purified from rat liver. (Uniprot: O95954)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10841
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.