Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5T4 Rabbit anti-Human, Mouse, Rat, Clone: 2K10S6, Novus Biologicals™
SDP

Catalog No. NB166624 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166624 20 μg
NB166623 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166624 Supplier Novus Biologicals Supplier No. NBP31576420UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

5T4 Monoclonal antibody specifically detects 5T4 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen 5T4
Applications Western Blot
Classification Monoclonal
Clone 2K10S6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 321-420 of human 5T4 (Q13641).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7162
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.