Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 6,8-Bis(benzylthio)octanoic acid
GREENER_CHOICE

Catalog No. p-7110817
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PDH and KGDH inhibitor

6,8-Bis(benzylthio)octanoic acid is a pyruvate dehydrogenase (PDH) and α-ketoglutarate dehydrogenase (KGDH) inhibitor. Disrupts tumor cell mitochondrial metabolism, and increases mitochondrial reactive oxygen species (ROS) production in H460 lung carcinoma cells. Has no effect on KGDH activity in normal bronchial epithelial cells. Induces cell death of multiple tumor cell lines selectively over normal cells in vitro. Inhibits tumor growth of H460 and BxPC-3 tumor cell xenografts in mice.

Specifications

Chemical Name or Material 6,8-Bis[(phenylmethyl)thio]octanoic acid
CAS 95809-78-2
Quantity 10 mg
Target Dehydrogenase Inhibitors
Molecular Formula C22H28O2S2
Formula Weight 388.59
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.