Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-1-methyl-5-(propylamino)uracil 98.0+%, TCI America™
SDP

Catalog No. A27151G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 125092-42-4
Molecular Formula C8H14N4O2
Molecular Weight (g/mol) 198.226
MDL Number MFCD01074825
InChI Key PXXPWRGFQVGQAZ-UHFFFAOYSA-N
PubChem CID 14406894
IUPAC Name 6-amino-1-methyl-5-(propylamino)pyrimidine-2,4-dione
SMILES CCCNC1=C(N(C(=O)NC1=O)C)N

Specifications

Melting Point 220°C
Color Yellow
Quantity 1 g
Formula Weight 198.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-methyl-5-(propylamino)uracil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.