Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Benzyloxypurine 98.0+%, TCI America™
SDP

Catalog No. B335525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B335525G 25 g
B33555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B335525G Supplier TCI America Supplier No. B335525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57500-07-9
Molecular Formula C12H10N4O
Molecular Weight (g/mol) 226.239
MDL Number MFCD00213666
InChI Key ZZZXGPGVDJDFCJ-UHFFFAOYSA-N
PubChem CID 853677
IUPAC Name 6-phenylmethoxy-7H-purine
SMILES C1=CC=C(C=C1)COC2=NC=NC3=C2NC=N3

Specifications

Melting Point 171°C
Quantity 25 g
Formula Weight 226.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Benzyloxypurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.