Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2-methyl-3-nitropyridine, 97%

Catalog No. AAH6429914
Click to view available options
Quantity:
5 g
25 g

CAS: 22282-96-8 | C6H5BrN2O2 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-96-8
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD03095219
InChI Key PCNVKBBKROTUNS-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro-6-picoline, 2-bromo-6-methyl-5-nitropyridine, 2-bromo-5-nitro-6-methylpyridine, 6-bromo-2-methyl-3-nitro-pyridine, 6-bromo-3-nitro-2-picoline, 6-bromo-3-nitropicoline, 6-brom-2-methyl-3-nitro-pyridin, pubchem2968, acmc-209fuh, ksc549a9p
PubChem CID 24728014
IUPAC Name 6-bromo-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(Br)=N1)[N+]([O-])=O

Specifications

Melting Point 67°C to 72°C
Quantity 25 g
Beilstein 135841
Formula Weight 217.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromo-2-methyl-3-nitropyridine
Hazard Category H302-H315-H318-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.