Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2-naphthyl Trifluoromethanesulfonate 95.0+%, TCI America™
SDP

Catalog No. T189425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T189425G 25 g
T18945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T189425G Supplier TCI America Supplier No. T189425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 151600-02-1
Molecular Formula C11H6BrF3O3S
Molecular Weight (g/mol) 355.13
MDL Number MFCD03093642
InChI Key FRNRXKFDZXFNKG-UHFFFAOYSA-N
Synonym 6-Bromo-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 6-Bromo-2-naphthyl Ester
PubChem CID 15311216
IUPAC Name 6-bromonaphthalen-2-yl trifluoromethanesulfonate
SMILES FC(F)(F)S(=O)(=O)OC1=CC=C2C=C(Br)C=CC2=C1

Specifications

Melting Point 54°C
Color Yellow
Quantity 25 g
Formula Weight 355.13
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-naphthyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.