Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-1-hexene, 97%

Catalog No. AAH5339614
Click to view available options
Quantity:
5 g
25 g

CAS: 928-89-2 | C6H11Cl | 118.604 g/mol

6-Chloro-1-hexene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-89-2
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.604
MDL Number MFCD00039391
InChI Key BLMIXWDJHNJWDT-UHFFFAOYSA-N
Synonym 6-chloro-1-hexene, 1-hexene, 6-chloro, 5-hexenyl chloride, 6-chlorohexene, chloro-1-hex, 6-chloro-1-hexen, 1-chloro-5-hexene, 1-chlorohex-5-ene, acmc-20alz4, 5-hexen-1-yl chloride
PubChem CID 70233
IUPAC Name 6-chlorohex-1-ene
SMILES C=CCCCCCl

Specifications

Density 0.896
Boiling Point 135°C to 136°C
Flash Point 29°C (84°F)
Refractive Index 1.438
Linear Formula Cl(CH2)4CH=CH2
Quantity 25 g
UN Number UN1993
Formula Weight 118.61
Percent Purity 97%
Chemical Name or Material 6-Chloro-1-hexene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-186-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.