Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloroguanine Crystalline MP Biomedicals

Catalog No. ICN19017901
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 10310-21-1
Molecular Formula C5H4ClN5
Molecular Weight (g/mol) 169.57
MDL Number MFCD00075252
InChI Key RYYIULNRIVUMTQ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine
PubChem CID 5360349
ChEBI CHEBI:72345
IUPAC Name 6-chloro-7H-purin-2-amine
SMILES NC1=NC(Cl)=C2NC=NC2=N1

Specifications

Quantity 1 g
Formula Weight 169.6
Physical Form Crystalline
Chemical Name or Material 6-Chloroguanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.