Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloromethyluracil 98.0+%, TCI America™
SDP

Catalog No. C105725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C105725G 25 g
C10575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C105725G Supplier TCI America Supplier No. C105725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18592-13-7
Molecular Formula C5H5ClN2O2
Molecular Weight (g/mol) 160.56
MDL Number MFCD00010095
InChI Key VCFXBAPEXBTNEA-UHFFFAOYSA-N
Synonym 6-chloromethyl uracil, 6-chloromethyl pyrimidine-2,4 1h,3h-dione, 6-chloromethyluracil, 2,4 1h,3h-pyrimidinedione, 6-chloromethyl, 6-chloromethyl-1h-pyrimidine-2,4-dione, 6-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 6-chloromethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 4-chloromethyl uracil, 6-chloromethyl-uracil, 6-chloromethyl uracil?
PubChem CID 73273
IUPAC Name 6-(chloromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES ClCC1=CC(=O)NC(=O)N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 160.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloromethyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.