Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloropurine, 99%

Catalog No. p-7081291
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 87-42-3 | C5H3ClN4 | 154.557 g/mol

6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

Solubility
Soluble in hot water and dimethyl formamide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-42-3
Molecular Formula C5H3ClN4
Molecular Weight (g/mol) 154.557
MDL Number MFCD00075825
InChI Key ZKBQDFAWXLTYKS-UHFFFAOYSA-N
Synonym 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
PubChem CID 5359277
IUPAC Name 6-chloro-7H-purine
SMILES C1=NC2=C(N1)C(=NC=N2)Cl

Specifications

Melting Point >300°C (decomposition)
Quantity 1 g
Beilstein 5774
Merck Index 14,2161
Solubility Information Soluble in hot water and dimethyl formamide.
Formula Weight 154.56
Percent Purity 99%
Chemical Name or Material 6-Chloropurine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-745-6
RTECSNumber UO7520000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.