Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloropurine, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
87-42-3
C5H3ClN4
154.56
MFCD00075825
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
6-chloropurine,6-chloro-9h-purine,1h-purine, 6-chloro,6-chloro-1h-purine,purine, 6-chloro,6-cp,6-chlorpurine,6-chloro purine,6-chloropurin,chloropurine, 6
5359277
6-chloro-7H-purine
C1=NC2=C(N1)C(=NC=N2)Cl
87-42-3
C5H3ClN4
154.56
MFCD00075825
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
6-chloropurine,6-chloro-9h-purine,1h-purine, 6-chloro,6-chloro-1h-purine,purine, 6-chloro,6-cp,6-chlorpurine,6-chloro purine,6-chloropurin,chloropurine, 6
5359277
6-chloro-7H-purine
C1=NC2=C(N1)C(=NC=N2)Cl
Melting Point >300°C
Sulfated Ash 0.1% max. (800°C)
Quantity 25g
Appearance (0.1% soln. in 0.1M HCl) clear, opalescent to yellowish
Elemental Analysis (on dry substance) :
Color Orange-Yellow to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Loss on Drying 0.5% max. (105°C, 5 hrs)
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Merck Index 15, 2163
Solubility Information Solubility in water: 5g/L water
Formula Weight 154.56
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material 6-chloropurine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-745-6